Learn More
Waters Corp Waters Corporation Autosampler Vial, LCGC Certified, Amber Glass, 12 x 32 mm, 2 mL
Catalog No. 50782525 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
LCGC certified amber glass screw neck vial designed for sensitive samples. Tested for HPLC cleanliness, ensuring no trace chemicals from manufacturing. Features robust design with tight dimensional tolerances for secure fit and chemical inertness for accurate analysis.
  • HPLC tested for cleanliness.
  • Adheres to stringent dimensional tolerances.
  • Amber glass protects light-sensitive samples.
  • Chemically inert and stable.
  • UV-testing detects trace chemicals.
  • Universal screw cap ensures secure seal.
  • Ideal for MS applications.
Catalog No. 50-782-525 Supplier Waters Corp Supplier No. 186000846C
LCGC certified amber glass screw neck vial designed for sensitive samples. Tested for HPLC cleanliness, ensuring no trace chemicals from manufacturing. Features robust design with tight dimensional tolerances for secure fit and chemical inertness for accurate analysis.
  • HPLC tested for cleanliness.
  • Adheres to stringent dimensional tolerances.
  • Amber glass protects light-sensitive samples.
  • Chemically inert and stable.
  • UV-testing detects trace chemicals.
  • Universal screw cap ensures secure seal.
  • Ideal for MS applications.