Learn More
Waters Corp Waters LCMS Certified Max Recovery Vial, 12 x 32 mm, 2 mL
Catalog No. 50782563 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This LCMS Certified Max Recovery Vial is designed for demanding LCMS applications, ensuring minimal sample loss and compatibility with all LCMS systems. It is constructed from clear glass and includes a cap with a preslit PTFE/silicone septum, making it suitable for multiple injections and a wide temperature range.
  • Max recovery vial design ensures minimal sample loss.
  • Compatible with all LCMS systems.
  • Withstands high temperatures and pressure.
  • Suitable for liquid and gas chromatography applications.
  • Rigorous quality control, certification, and high-quality materials.
  • Includes cap and preslit PTFE/silicone septum.
  • Preslit PTFE/silicone septa allow multiple injections, good resealing, and venting, with a working temperature range from -40°C to 200°C.
Catalog No. 50-782-563 Supplier Waters Corp Supplier No. 600000670CV
This LCMS Certified Max Recovery Vial is designed for demanding LCMS applications, ensuring minimal sample loss and compatibility with all LCMS systems. It is constructed from clear glass and includes a cap with a preslit PTFE/silicone septum, making it suitable for multiple injections and a wide temperature range.
  • Max recovery vial design ensures minimal sample loss.
  • Compatible with all LCMS systems.
  • Withstands high temperatures and pressure.
  • Suitable for liquid and gas chromatography applications.
  • Rigorous quality control, certification, and high-quality materials.
  • Includes cap and preslit PTFE/silicone septum.
  • Preslit PTFE/silicone septa allow multiple injections, good resealing, and venting, with a working temperature range from -40°C to 200°C.