Learn More
Waters Corp WATERS CORPORATION Screw Neck Vial, Polypropylene, 12 x 32 mm, 300 μL
Catalog No. 50781251 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This vial is a clean polypropylene-molded vial designed for applications where compound or molecule adherence to a polar glass surface is a concern. It offers a cost-effective alternative to traditional glass vials.
  • Polypropylene construction
  • Screw neck closure
  • 12 x 32 mm dimensions
  • 300 μL volume capacity
  • Ideal for autosampler use
  • Suitable for sample preparation
  • Packaged as 100 vials per unit
Catalog No. 50-781-251 Supplier Waters Corp Supplier No. 186002626
This vial is a clean polypropylene-molded vial designed for applications where compound or molecule adherence to a polar glass surface is a concern. It offers a cost-effective alternative to traditional glass vials.
  • Polypropylene construction
  • Screw neck closure
  • 12 x 32 mm dimensions
  • 300 μL volume capacity
  • Ideal for autosampler use
  • Suitable for sample preparation
  • Packaged as 100 vials per unit