Learn More
Waters Corp Waters Vial, Polypropylene, 12 x 32 mm, Snap Neck, 300 μL
Catalog No. 50781252 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This clean polypropylene-molded vial is a cost-effective alternative to glass vials, especially in applications where there is a concern that compounds or molecules may stick to a polar glass surface.
  • Clean polypropylene-molded construction
  • Cost-effective alternative to glass vials
  • Suitable for applications where compounds or molecules may stick to a polar glass surface
  • Has a volume capacity of 300 μL
  • Can be used for both liquid and solid sample storage
  • Compatible with most automated sample handling systems
  • Recommended for use within a temperature range of −40°C to 100°C
  • Easily sterilized using common laboratory methods as vials are not pre-sterilized
Catalog No. 50-781-252 Supplier Waters Corp Supplier No. 186002628
This clean polypropylene-molded vial is a cost-effective alternative to glass vials, especially in applications where there is a concern that compounds or molecules may stick to a polar glass surface.
  • Clean polypropylene-molded construction
  • Cost-effective alternative to glass vials
  • Suitable for applications where compounds or molecules may stick to a polar glass surface
  • Has a volume capacity of 300 μL
  • Can be used for both liquid and solid sample storage
  • Compatible with most automated sample handling systems
  • Recommended for use within a temperature range of −40°C to 100°C
  • Easily sterilized using common laboratory methods as vials are not pre-sterilized