Learn More
Waters Corp Waters Vial, Clear Glass, 12 x 32 mm, Snap Neck, 2 mL
Catalog No. 50818630 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This autosampler vial provides secure storage and retrieval of samples. Made from clear glass with 12 x 32 mm dimensions, it is compatible with most autosamplers. The snap neck ensures quick sealing, and the Max Recovery feature optimizes sample retrieval for precious samples. Manufactured with chemically resistant materials, these 2 mL vials (recommended 1.8 mL fill) are suitable for liquid and solid samples. Each pack contains 100 vials.
  • Clear glass construction
  • 12 x 32 mm dimensions
  • Snap neck design
  • Max recovery feature
  • 2 mL capacity (1.8 mL recommended fill)
  • Autosampler vial type
  • 100 vials per pack
  • Suitable for sample preparation
  • Chemically resistant materials
Catalog No. 50-818-630 Supplier Waters Corp Supplier No. 186000984
This autosampler vial provides secure storage and retrieval of samples. Made from clear glass with 12 x 32 mm dimensions, it is compatible with most autosamplers. The snap neck ensures quick sealing, and the Max Recovery feature optimizes sample retrieval for precious samples. Manufactured with chemically resistant materials, these 2 mL vials (recommended 1.8 mL fill) are suitable for liquid and solid samples. Each pack contains 100 vials.
  • Clear glass construction
  • 12 x 32 mm dimensions
  • Snap neck design
  • Max recovery feature
  • 2 mL capacity (1.8 mL recommended fill)
  • Autosampler vial type
  • 100 vials per pack
  • Suitable for sample preparation
  • Chemically resistant materials