Learn More
Microsolv Technology Corporation 12X32Mm, 2Ml Wide Opening Vial
Catalog No. NC1671300
Encompass_Preferred
Vials, Snap Top, Glass. Amber, 2ml, with Write-On Patch and fill lines. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. RSA Brand. 100/PK.
Catalog No. NC1671300 Supplier Microsolv Technology Corporation Supplier No. 9509CWAVRS
Vials, Snap Top, Glass. Amber, 2ml, with Write-On Patch and fill lines. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. RSA Brand. 100/PK.