Learn More
Waters Corp Waters Corporation Qsert Vial, Clear Glass, 12 x 32 mm, Snap Neck, 300 μL
Catalog No. 50780567 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Clear Glass Snap Neck Qsert Vial, with Cap and Preslit PTFE/Silicone Septum, ensures excellent sealing and HPLC compatibility via tight dimensional tolerances. The PTFE/Silicone Septum provides strong resealing and chemical resistance, maintaining silicone compatibility post-penetration. It operates from -40°C to 200°C and is made in facilities conforming to cGMP and ISO 9002 standards.
  • Excellent sealing mechanism from the cap and preslit PTFE/Silicone septum
  • Tight dimensional tolerances for compatibility with most HPLC systems
  • PTFE/Silicone septum offers great resealing properties and chemical resistance
  • Septum maintains silicone chemical compatibility after penetration
  • Wide working temperature range of -40°C to 200°C for the PTFE/Silicone septum
  • Manufactured in state-of-the-art facilities
  • Conforms to cGMP and ISO 9002 standards
Catalog No. 50-780-567 Supplier Waters Corp Supplier No. 186001125
This Clear Glass Snap Neck Qsert Vial, with Cap and Preslit PTFE/Silicone Septum, ensures excellent sealing and HPLC compatibility via tight dimensional tolerances. The PTFE/Silicone Septum provides strong resealing and chemical resistance, maintaining silicone compatibility post-penetration. It operates from -40°C to 200°C and is made in facilities conforming to cGMP and ISO 9002 standards.
  • Excellent sealing mechanism from the cap and preslit PTFE/Silicone septum
  • Tight dimensional tolerances for compatibility with most HPLC systems
  • PTFE/Silicone septum offers great resealing properties and chemical resistance
  • Septum maintains silicone chemical compatibility after penetration
  • Wide working temperature range of -40°C to 200°C for the PTFE/Silicone septum
  • Manufactured in state-of-the-art facilities
  • Conforms to cGMP and ISO 9002 standards