Learn More
Waters Corp Waters Corporation Qsert Vial, Clear Glass, 12 x 32 mm, 300 μL
Catalog No. 50780568 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters offers a selection of snap cap vials and snap caps/septa. This specific product is a Clear Glass 12 x 32 mm Snap Neck Qsert Vial, with Cap and PTFE Septum, 300 μL Volume, sold in packages of 100.
  • Includes a vial, cap, and septum
  • Snap neck closure type
  • Made of clear glass
  • PTFE seal material
  • Blue cap color
  • Dimensions: 12 x 32 mm
  • Volume capacity: 300 μL
  • Autosampler vial type
  • Designed for sample preparation
  • Package contains 100 units
Catalog No. 50-780-568 Supplier Waters Corp Supplier No. 186001127
Waters offers a selection of snap cap vials and snap caps/septa. This specific product is a Clear Glass 12 x 32 mm Snap Neck Qsert Vial, with Cap and PTFE Septum, 300 μL Volume, sold in packages of 100.
  • Includes a vial, cap, and septum
  • Snap neck closure type
  • Made of clear glass
  • PTFE seal material
  • Blue cap color
  • Dimensions: 12 x 32 mm
  • Volume capacity: 300 μL
  • Autosampler vial type
  • Designed for sample preparation
  • Package contains 100 units