Learn More
Waters Corp Waters Corporation Vial, LCMS Certified, Amber Glass, 12 x 32 mm, 2 mL
Supplier:  Waters Corp 600000752CV
Encompass_Preferred
This LCMS Certified Amber Glass 12 x 32 mm Screw Neck Vial prevents photosensitive samples from deteriorating. Its robust, inert composition withstands chemical and mechanical stresses, ensuring analysis integrity and a tight fit in analytical equipment. Rigorous testing detects contaminants. These vials meet stringent dimensional tolerances. A PTFE septum offers high solvent resistance. A universal screw cap provides a secure seal. All vials are certified.
  • Compatible with autosamplers.
  • Includes vial, cap, & septum.
Catalog No. 50-782-568