Learn More
Waters Corp WATERS CORPORATION Qsert Vial, Clear Glass, 12 x 32 mm, Screw Neck, 300 μL
Catalog No. 50781372 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Clear Glass 12 x 32 mm Screw Neck Qsert Vial is designed for various scientific applications, including sample preparation. It features a 300 μL volume capacity and is suitable for both liquid and gas samples.
  • Screw neck closure
  • Dimensions: 12 x 32 mm
  • Constructed from clear glass
  • 300 μL volume capacity
  • Designed as an autosampler vial
  • Includes vial
  • Withstands high-temperature applications
  • Compatible with automated sample handling systems
  • Suitable for both liquid and gas samples
  • Pre-cleaned and ready for use
Catalog No. 50-781-372 Supplier Waters Corp Supplier No. 186002804
The Clear Glass 12 x 32 mm Screw Neck Qsert Vial is designed for various scientific applications, including sample preparation. It features a 300 μL volume capacity and is suitable for both liquid and gas samples.
  • Screw neck closure
  • Dimensions: 12 x 32 mm
  • Constructed from clear glass
  • 300 μL volume capacity
  • Designed as an autosampler vial
  • Includes vial
  • Withstands high-temperature applications
  • Compatible with automated sample handling systems
  • Suitable for both liquid and gas samples
  • Pre-cleaned and ready for use