Learn More
Sarstedt Inc 12x75mm Tube, 5mL, Round Bottom, 1000/Pack
Catalog No. 50809214 Shop All Sarstedt Inc. Products
Encompass_Preferred
12x75mm Tube 5mL Round Bottom Polypropylene No Cap
Catalog No. 50-809-214 Supplier Sarstedt Inc Supplier No. 55.526.305
12x75mm Tube 5mL Round Bottom Polypropylene No Cap