Learn More
MSC Baldwin Outer Air Filter, 9.219 in. OD, 1 ea
Catalog No. 5000505665
Encompass
This outer air filter element is designed for use in agricultural equipment, providing essential air filtration for various machinery.
  • Element style
  • Air filter type
  • Inside diameter 0.656/4.844 in
  • Length 13.375 in
  • Outside diameter 9.219 in
  • Replaces Case-IH 87682983 OEM part
Catalog No. 50-005-05665 Supplier MSC Supplier No. 10940211
This outer air filter element is designed for use in agricultural equipment, providing essential air filtration for various machinery.
  • Element style
  • Air filter type
  • Inside diameter 0.656/4.844 in
  • Length 13.375 in
  • Outside diameter 9.219 in
  • Replaces Case-IH 87682983 OEM part