Learn More
Medchemexpress LLC 13,13-dimethyl-11-oxo-4,7,12-trioxa-10-azatetradecanoic acid | 1365655-91-9 | MFCD22056304 | 98.0% | 277.31 g/mol | C12H23NO6 | 25 G
SDP
Catalog No. 5000303684
Encompass_Preferred
Boc-NH-PEG2-CH2CH2COOH is a Boc-protected PEG2 linker with a terminal carboxylic acid used for constructing PROTACs and other small-molecule conjugates. It offers a short, hydrophilic spacer and a protected amine for selective coupling and downstream deprotection.
  • Boc-protected amino group for orthogonal protection.
  • Terminal carboxylic acid for standard amide coupling.
  • Short PEG2 spacer increases solubility and conformational flexibility.
  • High purity suitable for research synthesis and derivatization.
  • Liquid form and defined density facilitate solution preparation.
Catalog No. 50-003-03684 Supplier Medchemexpress LLC Supplier No. 25GHYW025812
Boc-NH-PEG2-CH2CH2COOH is a Boc-protected PEG2 linker with a terminal carboxylic acid used for constructing PROTACs and other small-molecule conjugates. It offers a short, hydrophilic spacer and a protected amine for selective coupling and downstream deprotection.
  • Boc-protected amino group for orthogonal protection.
  • Terminal carboxylic acid for standard amide coupling.
  • Short PEG2 spacer increases solubility and conformational flexibility.
  • High purity suitable for research synthesis and derivatization.
  • Liquid form and defined density facilitate solution preparation.