Learn More
Medchemexpress LLC 13,14-Dihydro-15-keto-PGE2-d9 | 2750534-81-5 | 361.52 | 25 UG
SDP
Catalog No. 5000439445
Encompass_Preferred
13,14-Dihydro-15-keto-PGE2-d9 is the deuterium labeled 13,14-Dihydro-15-keto-PGE2. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements
  • Affects pharmacokinetic and metabolic profiles of drugs
  • For research use only
Catalog No. 50-004-39445 Supplier Medchemexpress LLC Supplier No. 25UGHY113254S1
13,14-Dihydro-15-keto-PGE2-d9 is the deuterium labeled 13,14-Dihydro-15-keto-PGE2. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements
  • Affects pharmacokinetic and metabolic profiles of drugs
  • For research use only