Learn More
Cayman Chemical 13,14-DIHYDRO-15-KETO PROS 1MG
SDP
Supplier:  Cayman Chemical 136501
Encompass_Preferred
13,14-DIHYDRO-15-KETO PROSTAGLANDIN E1, ≥98% 1mg CAS#5094-14-4 C20H34O5 MW:354.5
Catalog No. NC1956345