Learn More
Enzo Life Sciences 13,14-Dihydro-prostaglandin E1 (1mg). CAS: 19313-28-1
SDP
Supplier:  Enzo Life Sciences BMLPG0480001
Encompass_Preferred
Circulating metabolite of PGE1 in man. Vasodilator equipotent with PGE12. Inhibits human platelet aggregation. Antimitotic activity. Alternative name: 13,14-Dihydro-PGE1. Purity: ≥98% (TLC). Long Term Storage: -80°C.
Catalog No. 50-201-1422