Learn More
Cayman Chemical 13,14-dihydro Prostaglandin F2α, Purity: ≥98%, CAS: 27376-74-5, 10 mg
SDP
Catalog No. 502965576
Encompass_Preferred
An analog of PGF2α which has no unsaturation in the lower side chain; causes luteolysis in hamsters with a potency five times higher than PGF2α
Catalog No. 50-296-5576 Supplier Cayman Chemical Supplier No. 1666010 MG
An analog of PGF2α which has no unsaturation in the lower side chain; causes luteolysis in hamsters with a potency five times higher than PGF2α