Learn More
Cayman Chemical 13 14dIhydro15ketotetrno 10ug
SDP
Catalog No. 502954708
Encompass_Preferred
A potential metabolite of PGD2, presumably produced from the known metabolite 13,14-dihydro-15-keto PGD2
Catalog No. 50-295-4708 Supplier Cayman Chemical Supplier No. 1310010 UG
A potential metabolite of PGD2, presumably produced from the known metabolite 13,14-dihydro-15-keto PGD2