Learn More
Ace Glass, Inc. 13.2L solution bottle plastic coated
SDP
Catalog No. 50971857
Encompass_Preferred
13.2L solution bottle is coated to the neck with PVC safety coating to reduce potential breakage, and exposure to laboratory personnel when handling hazardous material. Carboy style bottles with sloping shoulders. Necks are tooled for uniform fit with No. 12 stoppers (not supplied).
Catalog No. 50-971-857 Supplier Ace Glass, Inc. Supplier No. 539307
13.2L solution bottle is coated to the neck with PVC safety coating to reduce potential breakage, and exposure to laboratory personnel when handling hazardous material. Carboy style bottles with sloping shoulders. Necks are tooled for uniform fit with No. 12 stoppers (not supplied).