Learn More
Cold Chain Technologies Frozen Shipper, Available Payload Dimensions 3 3/8 X 9 1/2 X 6, System Weight 70.7 lbs, Outside Dimensions 22 7/8 x 19 3/8 x 19 3/8
Catalog No. NC1475860
Encompass_Preferred
This item has a minimum qty of 4 per supplier requirements.
Utilizes dry ice pellets, 2 size parcel containers 3.5L & 12.5L Payload Capacities, Superior pre-qualified temperature protection for deep frozen (below -70C) applications for 120+ hours, Designed to maintain temperature no matter the orientation of the box during transit. Includes payload and positioning boxes (for dry ice sublimation) to ensure full product protection and complete coverage of dry ice during shipment.
Catalog No. NC1475860 Supplier Cold Chain Technologies Supplier No. STSU186DIULSU
This item has a minimum qty of 4 per supplier requirements.
Utilizes dry ice pellets, 2 size parcel containers 3.5L & 12.5L Payload Capacities, Superior pre-qualified temperature protection for deep frozen (below -70C) applications for 120+ hours, Designed to maintain temperature no matter the orientation of the box during transit. Includes payload and positioning boxes (for dry ice sublimation) to ensure full product protection and complete coverage of dry ice during shipment.