Learn More
AdipoGen 13-cis-4-Oxoretinoic acid
SDP
Catalog No. 502560325
Encompass_Preferred
Chemical. CAS 71748-58-8. Formula C20H26O3. MW 314.42. Synthetic. Mayor active metabolites of 13-cis-retinoic acid important drug in dermatology and treatment of neuroblastoma with effects on gene transcription that regulate growth, differentiation and inflammation in normal and neoplastic skin cells. Shown to be as active as 13-cis-retinoic acid against neuroblastoma cell lines. Can be used as reference compound.
Catalog No. 50-256-0325 Supplier AdipoGen Supplier No. CDXO0048M025
Chemical. CAS 71748-58-8. Formula C20H26O3. MW 314.42. Synthetic. Mayor active metabolites of 13-cis-retinoic acid important drug in dermatology and treatment of neuroblastoma with effects on gene transcription that regulate growth, differentiation and inflammation in normal and neoplastic skin cells. Shown to be as active as 13-cis-retinoic acid against neuroblastoma cell lines. Can be used as reference compound.