Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

13-cis-Retinal, Thermo Scientific™

Catalog No. AC418955000
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC418955000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC418955000 Supplier Thermo Scientific Chemicals Supplier No. 418955000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 472-86-6
Molecular Formula C20H28O
Molecular Weight (g/mol) 284.44
MDL Number MFCD00074858,MFCD00001550
InChI Key NCYCYZXNIZJOKI-HWCYFHEPSA-N
Synonym 13-cis-retinal, 13-cis-retinaldehyde, neoretinene a, 13z-retinal, neovitamin a aldehyde, 13-cis retinal, unii-ibp64540cd, retinal, 13-cis, 13-cis-vitamin a aldehyde
PubChem CID 6436079
ChEBI CHEBI:45487
IUPAC Name (2Z,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenal
SMILES C\C(\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C)=C\C=O

Specifications

Quantity 500 mg
Formula Weight 284.44
Chemical Name or Material 13-cis-Retinal
Hazard Category PRODUCT NEEDS INERT ATMOSPHERE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.