Learn More
Enzo Life Sciences 13-cis-Retinoic acid (500mg). CAS: 4759-48-2
SDP
Supplier:  Enzo Life Sciences BMLGR1020500
Encompass_Preferred
A potent modulator of growth and differentiation. Clinically used in a variety of skin disorders. Alternative name: Isotretinoin. Purity: ≥95% (TLC). Solubility: Soluble in DMSO (25mg/ml) or ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0914