Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 13-cis-Retinyl acetate | 34356-31-5 | 85.0% | C22H32O2 | 1 MG
SDP

Catalog No. 5000356168
Encompass_Preferred

13-cis-Retinyl acetate is a 13-cis isomer formed by Retinyl acetate and is an active metabolite of vitamin A, intended for research use only.

  • Active metabolite of vitamin A
  • For research use only
  • Molecular weight: 328.49
  • Appearance: oil
  • Color: off-white to light yellow
  • Endogenous metabolite

Catalog No. 50-003-56168 Supplier Medchemexpress LLC Supplier No. HY1458061MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

13-cis-Retinyl acetate is a 13-cis isomer formed by Retinyl acetate and is an active metabolite of vitamin A, intended for research use only.

  • Active metabolite of vitamin A
  • For research use only
  • Molecular weight: 328.49
  • Appearance: oil
  • Color: off-white to light yellow
  • Endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.