Learn More
Sigma Aldrich Fine Chemicals Biosciences 13-Dimethoxybenzene 98 fG1KG
Catalog No. 502899491
Encompass
1 3-Dimethoxybenzene can be used as a flavoring agent in the food industry. It is reported to be found in the volatiles of port wine and Roquefort cheese.
Catalog No. 50-289-9491 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W2385031KGK
1 3-Dimethoxybenzene can be used as a flavoring agent in the food industry. It is reported to be found in the volatiles of port wine and Roquefort cheese.