Learn More
MCR Safety Cut Pro™ 9693 Aramid Fiber Gloves, Nitrile Coated
Catalog No. 191304021D Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-4021D X-Large
19-130-4021A Small
19-130-4021B Medium
19-130-4021C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-130-4021D Supplier MCR Safety Supplier No. 9693XL
This item is not returnable. View return policy
Gloves constructed of yellow aramid fiber shell feature a black textured nitrile coating on the palm and fingertips for gripping power in wet or dry conditions.
  • 15-gauge aramid fiber shell
  • Black textured nitrile palm
  • Seamless knit
  • Allows manual dexterity
  • Durable and comfortable
Specifications
ANSI Abrasion Rating ANSI 3
Certifications/Compliance ASTM F1060-18
Construction Seamless Knit
Coating Material Nitrile
Color Black, Yellow
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free Yes
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size X-Large
For Use With (Application) Abrasive applications, assembly, box handling, brick handling, construction, cut protection, farming, general maintenance, knife handling, landscaping, plumbing, recycling
Glove Cuff Style Knit
Hem Color White
Model 9693XL
Series Cut Pro™ 9693
Yarn Gauge 15