Learn More
Cayman Chemical 13-methyl Myristic Acid, Purity: ≥98%, CAS: 2485-71-4, 25 mg
SDP
Catalog No. 502932549
Encompass_Preferred
A methylated fatty acid; has been found in dairy ewe rumen liquor, bovine milk, goat milk fat, the edible flesh of rainbow trout, and Flavobacterium; levels decrease in arable soil in response to heavy metal contamination
Catalog No. 50-293-2549 Supplier Cayman Chemical Supplier No. 2481125 MG
A methylated fatty acid; has been found in dairy ewe rumen liquor, bovine milk, goat milk fat, the edible flesh of rainbow trout, and Flavobacterium; levels decrease in arable soil in response to heavy metal contamination