Learn More
Cayman Chemical 13-methyl MyrIstIc AcId 5mg
SDP
Catalog No. 502934210
Encompass_Preferred
A methylated fatty acid; has been found in dairy ewe rumen liquor, bovine milk, goat milk fat, the edible flesh of rainbow trout, and Flavobacterium; levels decrease in arable soil in response to heavy metal contamination
Catalog No. 50-293-4210 Supplier Cayman Chemical Supplier No. 248115 MG
A methylated fatty acid; has been found in dairy ewe rumen liquor, bovine milk, goat milk fat, the edible flesh of rainbow trout, and Flavobacterium; levels decrease in arable soil in response to heavy metal contamination