Learn More
Cayman Chemical 13-Methylberberine (chloride), Purity: ≥98%, CAS: 54260-72-9, 5 mg
SDP
Catalog No. 502983609
Encompass_Preferred
A 13-methyl-substituted derivative of berberine; has antibacterial, anti-inflammatory, anti-obesity, and hypercholesterolemic activity; has improved antibacterial activity against S. aureus (MIC = 125 µg/ml) and improved antitumor activity compared to berberine; induces down-regulation of adipocyte differentiation transcription factors; reduces de novo lipid synthesis
Catalog No. 50-298-3609 Supplier Cayman Chemical Supplier No. 211545 MG
A 13-methyl-substituted derivative of berberine; has antibacterial, anti-inflammatory, anti-obesity, and hypercholesterolemic activity; has improved antibacterial activity against S. aureus (MIC = 125 µg/ml) and improved antitumor activity compared to berberine; induces down-regulation of adipocyte differentiation transcription factors; reduces de novo lipid synthesis