Learn More
Savillex Corporation 13 mL PFA Vial, Flat Interior
SDP
Catalog No. NC1673900
Encompass
Fits other autosamplers used in ICP-MS and GFAAS. Flat interior base. Service temperature range of PFA is ‑328°F to 500°F (‑200°C to 260°C).
Catalog No. NC1673900 Supplier Savillex Corporation Supplier No. 20001314
Fits other autosamplers used in ICP-MS and GFAAS. Flat interior base. Service temperature range of PFA is ‑328°F to 500°F (‑200°C to 260°C).