Learn More
Cole-Parmer Polycarbonate Filter Holder, 13 mm, 12/pk
Supplier:  Cole-Parmer 2955040
Encompass_Preferred
These reusable holders are made of clear, autoclavable polycarbonate with silicone gasket or O-rings. The bell-shaped base protects the filtrate from repeated contamination while following in a receiver. Maximum operating pressure is 101 psi.
Catalog No. NC9461377