Learn More
Perkin Elmer US LLC Evacuable KBr Die
Catalog No. 509041593
Encompass_Preferred
The 15 Ton Hydraulic KBr Press is an easy-to-operate benchtop press. It products the highest quality KBr pellets, 13 mm in diameters.
Catalog No. 50-904-1593 Supplier Perkin Elmer US LLC Supplier No. 51001144
The 15 Ton Hydraulic KBr Press is an easy-to-operate benchtop press. It products the highest quality KBr pellets, 13 mm in diameters.