Learn More
Waters Corp Cap, 13 mm, Solid Black, PTFE/Silicone Liner, 100 pk
Catalog No. 502392701 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This solid black cap features high-quality plastic for durability and chemical resistance. Its PTFE (polytetrafluoroethylene) with silicone layer liner ensures enhanced sealing. Designed to securely seal both liquid and solid samples, it provides an excellent barrier against leakage and contamination. It is suitable for various sample types and fits standard laboratory vials with a 13 mm neck size.
  • Closure type: Screw neck
  • Cap color: Black
  • Seal material: PTFE/Silicone liner
  • Includes: Solid cap and liner
  • Units per package: 100 pk
  • Compatibility: Designed to fit standard laboratory vials with a 13 mm neck size
  • Autoclavable for sterilization purposes
  • High-quality plastic material ensures durability and chemical resistance
  • Provides an excellent barrier against leakage and contamination for both liquid and solid samples
Catalog No. 50-239-2701 Supplier Waters Corp Supplier No. 186007224
This solid black cap features high-quality plastic for durability and chemical resistance. Its PTFE (polytetrafluoroethylene) with silicone layer liner ensures enhanced sealing. Designed to securely seal both liquid and solid samples, it provides an excellent barrier against leakage and contamination. It is suitable for various sample types and fits standard laboratory vials with a 13 mm neck size.
  • Closure type: Screw neck
  • Cap color: Black
  • Seal material: PTFE/Silicone liner
  • Includes: Solid cap and liner
  • Units per package: 100 pk
  • Compatibility: Designed to fit standard laboratory vials with a 13 mm neck size
  • Autoclavable for sterilization purposes
  • High-quality plastic material ensures durability and chemical resistance
  • Provides an excellent barrier against leakage and contamination for both liquid and solid samples