Learn More
Cayman Chemical 13-OxoDE-d3 100ug
SDP
Catalog No. 502950745
Encompass_Preferred
An internal standard for the quantification of 13-OxoODE by GC- or LC-MS
Catalog No. 50-295-0745 Supplier Cayman Chemical Supplier No. 338620100 UG
An internal standard for the quantification of 13-OxoODE by GC- or LC-MS