Learn More
TRANSMED COMPANY LLC Drug Test Cup, 13 Panel, Click Cap, Lowere Cut-offs, AMP300, BUP, BZO, COC150, MDMA500, OPI300, OXY, THC, ETG500, FEN, K2, TRA, EDDP, 25/BX USE ONLY)
SDP
Catalog No. 501905727
Encompass_Preferred
Multi-Panel Drug Test Cup, 13 Panel, Click Cap, AMP300, BUP, BZO, COC150, MDMA500, OPI300, OXY, THC, ETG500, FEN, K2, TRA, EDDP, 25/BX (FORENSIC USE ONLY)
This item is not returnable. View return policy
Catalog No. 50-190-5727 Supplier TRANSMED COMPANY LLC Supplier No. HCDOAV4135SEF1KT
Multi-Panel Drug Test Cup, 13 Panel, Click Cap, AMP300, BUP, BZO, COC150, MDMA500, OPI300, OXY, THC, ETG500, FEN, K2, TRA, EDDP, 25/BX (FORENSIC USE ONLY)