Learn More
Cole-Parmer Reversible 13-Point Horizontal Temperature Label, 24-36C/75-97F; 10/Pk
Catalog No. NC1287640 Shop All Cole-Parmer Products
Encompass_Preferred
These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring temperatures. Indicating strips use liquid crystal technology to display the current temperature at an economical cost. The strip will turns a green color when ambient temperature is reached. Shelf life: 1 year after date of purchase.
Catalog No. NC1287640 Supplier Cole-Parmer Supplier No. 0903546
These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring temperatures. Indicating strips use liquid crystal technology to display the current temperature at an economical cost. The strip will turns a green color when ambient temperature is reached. Shelf life: 1 year after date of purchase.