Learn More
Cayman Chemical 13 RHODE cholesTRyl esTR 100ug
SDP
Supplier:  Cayman Chemical 38606100 UG
Encompass_Preferred
13(R)-HODE cholesteryl ester was originally extracted from atherosclerotic lesions.{2227} It remains uncertain whether the oxidized fatty acid portion of the molecule results from enzymatic lipoxygenation or from random lipid peroxidation.{1126} 13(R)-HODE cholesteryl ester can be used as a standard for analysis of chiral HODE cholesteryl esters.
Catalog No. 50-291-9043