Learn More
Cayman Chemical 13 S-HODE-d4 100ug
SDP
Catalog No. 502947139
Encompass_Preferred
An internal standard for the quantification of 13-HODE by GC- or LC-MS
Catalog No. 50-294-7139 Supplier Cayman Chemical Supplier No. 338610100 UG
An internal standard for the quantification of 13-HODE by GC- or LC-MS