Learn More
Cayman Chemical 13 SHODE cholesTRyl esTR 100ug
SDP
Catalog No. 502918984
Encompass_Preferred
13(S)-HODE cholesteryl ester was originally extracted from atherosclerotic lesions.{2227} It remains uncertain whether the oxidized fatty acid portion of the molecule results from enzymatic lipoxygenation or from random lipid peroxidation.{1126} 13(S)-HODE cholesteryl ester can be used as a standard for analysis of chiral HODE cholesteryl esters.
Catalog No. 50-291-8984 Supplier Cayman Chemical Supplier No. 38611100 UG
13(S)-HODE cholesteryl ester was originally extracted from atherosclerotic lesions.{2227} It remains uncertain whether the oxidized fatty acid portion of the molecule results from enzymatic lipoxygenation or from random lipid peroxidation.{1126} 13(S)-HODE cholesteryl ester can be used as a standard for analysis of chiral HODE cholesteryl esters.