Learn More
Chemglass Life Sciences #22 Outer Joint, 22mm X 1.8W Tubing
SDP
Supplier:  Chemglass Life Sciences CGB12505
Encompass_Preferred
#22 Outer Joint, 22mm X 1.8W Tubing. Standard taper flask length outer joints. Part number CGB-125-09 is used as a replacement neck for 2L Squibb style separatory funnels.
Catalog No. 50-153-1920