Learn More
Chemglass Life Sciences #38 Outer Joint, 45mm X 2.0W Tubing
SDP
Catalog No. 501531922
Encompass_Preferred
#38 Outer Joint, 45mm X 2.0W Tubing. Standard taper flask length outer joints. Part number CGB-125-09 is used as a replacement neck for 2L Squibb style separatory funnels.
Catalog No. 50-153-1922 Supplier Chemglass Life Sciences Supplier No. CGB12509
#38 Outer Joint, 45mm X 2.0W Tubing. Standard taper flask length outer joints. Part number CGB-125-09 is used as a replacement neck for 2L Squibb style separatory funnels.