Learn More
Chemglass Life Sciences #13 Outer Joint, 15mm X 1.2W Tubing
SDP
Catalog No. 501531917
Encompass_Preferred
#13 Outer Joint, 15mm X 1.2W Tubing. Standard taper flask length outer joints. Part number CGB-125-09 is used as a replacement neck for 2L Squibb style separatory funnels.
Catalog No. 50-153-1917 Supplier Chemglass Life Sciences Supplier No. CGB12502
#13 Outer Joint, 15mm X 1.2W Tubing. Standard taper flask length outer joints. Part number CGB-125-09 is used as a replacement neck for 2L Squibb style separatory funnels.