Learn More
Cayman Chemical 13(Z)-Docosenoic Acid, Purity: ≥98%, CAS: 112-86-7, 250 mg
SDP
Catalog No. 502895393
Encompass_Preferred
13(Z)-Docosenoic acid is a 22-carbon monounsaturated fatty acid. It is found predominantly in canola oil.{8144} 13(Z)-Docosenoic acid is metabolized to oleic acid in vivo. Diets rich in 13(Z)-docosenoic acid were shown to cause heart lipidosis in experimental animals.{7786,8145} The C-1 amide of docosenoic acid has been identified as one of the anandamide-related neurotransmitters associated with sleep.{1317}
Catalog No. 50-289-5393 Supplier Cayman Chemical Supplier No. 90175250 MG
13(Z)-Docosenoic acid is a 22-carbon monounsaturated fatty acid. It is found predominantly in canola oil.{8144} 13(Z)-Docosenoic acid is metabolized to oleic acid in vivo. Diets rich in 13(Z)-docosenoic acid were shown to cause heart lipidosis in experimental animals.{7786,8145} The C-1 amide of docosenoic acid has been identified as one of the anandamide-related neurotransmitters associated with sleep.{1317}