Learn More
Cayman Chemical 13(Z)-Docosenoic Acid (sodium salt), Purity: ≥98%, CAS: 14488-85-8, 10 mg
SDP
Catalog No. 502916252
Encompass_Preferred
A 22-carbon monounsaturated fatty acid; increases weight gain and heart triglyceride levels in calcium-deprived weanling rats in the diet at 6.6% w/w
Catalog No. 50-291-6252 Supplier Cayman Chemical Supplier No. 3897510 MG
A 22-carbon monounsaturated fatty acid; increases weight gain and heart triglyceride levels in calcium-deprived weanling rats in the diet at 6.6% w/w