Learn More
Cayman Chemical 13 Z-EIcosenoIc AcId 10mg
SDP
Catalog No. 502935064
Encompass_Preferred
An ω-7 fatty acid; increases triglyceride accumulation in 3T3-L1 cells at 50 μM
Catalog No. 50-293-5064 Supplier Cayman Chemical Supplier No. 2616910 MG
An ω-7 fatty acid; increases triglyceride accumulation in 3T3-L1 cells at 50 μM