Learn More
Cayman Chemical 13(Z)-Eicosenoic Acid, Purity: ≥95%, CAS: 17735-94-3, 25 mg
SDP
Catalog No. 502935086
Encompass_Preferred
An ω-7 fatty acid; increases triglyceride accumulation in 3T3-L1 cells at 50 μM
Catalog No. 50-293-5086 Supplier Cayman Chemical Supplier No. 2616925 MG
An ω-7 fatty acid; increases triglyceride accumulation in 3T3-L1 cells at 50 μM