Learn More
Cayman Chemical 13(Z)-Docosenoic Acid methyl ester, Purity: ≥95%, CAS: 1120-34-9, 500 mg
SDP
Catalog No. 502984850
Encompass_Preferred
A fatty acid methyl ester; a flavor-active, volatile, and aromatic compound found in cooked commercial shrimp waste; a component of biodiesel formed from C. megalocarpus and C. pentandra oils; has been used as a standard for the quantification of 13(Z)-docosenoic acid by GC-MS
Catalog No. 50-298-4850 Supplier Cayman Chemical Supplier No. 20568500 MG
A fatty acid methyl ester; a flavor-active, volatile, and aromatic compound found in cooked commercial shrimp waste; a component of biodiesel formed from C. megalocarpus and C. pentandra oils; has been used as a standard for the quantification of 13(Z)-docosenoic acid by GC-MS