Learn More
Sino Biological Recombinant Human P-Selectin/CD62P Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 13025H08H100
Encompass_Preferred
A DNA sequence encoding the human SELP (AAN06828.1) extracellular domain (Met 1-Ala 771) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8174