Learn More
Sino Biological Recombinant Human IGF-II Protein
Supplier:  Sino Biological 13032HNAY100
Encompass_Preferred
A DNA sequence encoding the human IGF2 (P01344-1) (Ala25-Glu91) was expressed.
Catalog No. 50-161-8187