Learn More
Sino Biological Recombinant Human TRF-1 Protein (His Tag)
Supplier:  Sino Biological 13044H07B50
Encompass_Preferred
A DNA sequence encoding the human TERF1 isoform 2 (NP_003209.2) (Met 1-Asp 419) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-8192